Learn More
eGFP Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-06640APC
Product Code. 18618737
  • 529.00 € / 0.10mL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
eGFP Polyclonal antibody specifically detects eGFP in Nonspecies specific samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
eGFP
Polyclonal
ELISA
Enhanced Green Fluorescent Protein, GFP, Green Fluorescent Protein
Produced in rabbits immunized with purified, recombinant eGFP (AFI57842.2; Met1-Lys239).
0.1 mL
Primary
Store at 4C in the dark.
IgG
ELISA
APC
PBS
Rabbit
Antigen and protein A Affinity-purified
RUO
Non-species specific
Purified
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions