Learn More
Human Carbohydrate Sulfotransferase 15/CHST15 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3365
This item is not returnable.
View return policy
Description
Carbohydrate Sulfotransferase 15/CHST15 Polyclonal specifically detects Carbohydrate Sulfotransferase 15/CHST15 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| Carbohydrate Sulfotransferase 15/CHST15 | |
| Polyclonal | |
| Unconjugated | |
| Q7LFX5 | |
| CHST15 | |
| Mouse myeloma cell line NS0-derived recombinant human Carbohydrate Sulfotransferase 15/CHST15 Ser99-Thr561 Accession # Q7LFX5 | |
| 100 μg | |
| Primary | |
| Detects human Carbohydrate Sulfotransferase 15/CHST15 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| B cell RAG associated protein (GALNAC4S-6ST), B-cell RAG-associated gene protein, BRAG, BRAGKIAA0598RP11-47G11.1, carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15, carbohydrate sulfotransferase 15, CHST15, DKFZp781H1369, EC 2.8.2.33, GalNAc4S-6ST, GALNAC4S6ST, hBRAG, MGC34346, N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 51363 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction