Learn More
GA International Laser and Inkjet Matte Paper Labels
Product Code. 17895076
Encompass_Preferred
Click to view available options
Dimensions (LxW):
100 x 30 mm
100.1 x 30 mm
105 x 37 mm
105 x 42.3 mm
105 x 74 mm
139 x 99.1 mm
199.6 x 143.5 mm
199.6 x 289 mm
200 x 60 mm
210 x 99 mm
31 x 22 mm
31.5 x 13 mm
45.7 x 25.4 mm
46 x 11.1 mm
47.5 x 21.2 mm
63.5 x 33.8 mm
63.5 x 38.1 mm
63.5 x 46.5 mm
63.5 x 72 mm
64 x 7 mm
70 x 36 mm
76.2 x 46.4 mm
99.1 x 34 mm
99.1 x 38.1 mm
99.1 x 57 mm
99.1 x 67.7 mm
99.1 x 93.1 mm
Unit Size:
Pack of 100
Pack of 1000
Pack of 1200
Pack of 12000
Pack of 12600
Pack of 1400
Pack of 1600
Pack of 1800
Pack of 200
Pack of 2100
Pack of 2400
Pack of 300
Pack of 400
Pack of 4000
Pack of 4800
Pack of 600
Pack of 7200
Pack of 8400
This item is not returnable. View return policy
Product Code. 17895076
Brand: GA International A4LIP124WH
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
These A4 labels are compatible with both desktop laser and inkjet printers and are ideal for general use applications in labs, offices, and warehouses.
  • Compatible with both laser and inkjet printers, for convenient printing
  • High print quality with vivid colors
  • Premium materials allow for uniform ink absorption and brightness
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Sheet
Material Thermoplastic Film
Printable Sides Single-side Printable
Print Technology Laser
Product Type Label
Sterility Non-sterile
Labels per Sheet 12/Sheet
No. per Pack 100/Pk.
Quantity 1200/Pk.
Shape Rectangle
Temperature Range (English) -20.2°F to +248°F
Temperature Range (Metric) -29°C to +120°C
Dimensions (LxW) 76.2 x 46.4 mm
Length (English) 1.83 in.
Length (Metric) 46.4 mm
Width (English) 3 in.
Width (Metric) 76.2 mm
Show More Show Less
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.