Learn More
Kautex™ LDPE Wide Mouth Bottles Series 303
Beschreibung
- Manufactured from low density polyethylene (LDPE) for flexibility and toughness
- Natural translucent color, round bottles
- Easy to fill and clean
- Ideal for dispensing, sending samples, adhesives, transportation and storage
- Includes leakproof screw closure
Spezifikation
Spezifikation
| Capacity (Metric) | 300 mL |
| For Use With (Application) | Dispensing, Sending Samples, Mailing, Transport, Storage |
| Form | Round |
| Includes | Closure |
| Material | LDPE |
| Neck ID (Metric) | 30.8 mm |
| Replacement Closure | Screw Closure LDPE |
| Quantity | 1 Ea. |
| Graduated | No |
| Height (Metric) | 128 mm |
| Mehr anzeigen |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?