Learn More
CAY10602, MedChemExpress
Product Code. 16494966
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy
Product Code. 16494966
Brand: Medchem Express HY104073
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
MedChemExpress CAY10602 is a SIRT1 activator. CAY10602 dose-dependently suppresses the NF-κB-dependent induction of TNF-α by lipopolysaccharide in THP-1 cells.
CAY10602 is a SIRT1 activator. CAY10602 dose-dependently suppresses the NF-κB-dependent induction of TNF-α by lipopolysaccharide in THP-1 cells.
Specifications
Chemical Name or Material CAY10602
CAS 374922-43-7
Color Gray
Purity Grade Notes Research
Molecular Formula C22H15FN4O2S
Quantity 100 mg
Solubility Information DMSO : 41.67 mg/mL (99.58 mM; Need ultrasonic)
SMILES NC1=C(S(=O)(C2=CC=CC=C2)=O)C3=NC4=CC=CC=C4N=C3N1C5=CC=C(F)C=C5
Molecular Weight (g/mol) 418.44
Formula Weight 418.44
Percent Purity 98.63%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.