Learn More
BI-9564, MedChemExpress
Product Code. 16480906
Encompass
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
2mg
50mg
5mg
This item is not returnable. View return policy
Product Code. 16480906
Brand: Medchem Express HY100352
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
MedChemExpress BI-9564 is a potent, selective and cell-permeable BRD9/BRD7 bromodomains inhibitor, with IC50s of 75 nM and 3.4 μM and Kds of 14 nM and 239 nM, respectively. BI-9564 has an IC50 of > 100 μM for BET family.
BI-9564 is a potent, selective and cell-permeable BRD9/BRD7 bromodomains inhibitor, with IC50s of 75 nM and 3.4 μM and Kds of 14 nM and 239 nM, respectively. BI-9564 has an IC50 of > 100 μM for BET family.
Specifications
Chemical Name or Material BI-9564
CAS 1883429-22-8
Color White
Purity Grade Notes Research
Molecular Formula C20H23N3O3
Quantity 200 mg
Solubility Information DMSO : 8.33 mg/mL (23.57 mM; Need ultrasonic)
SMILES CN(C)CC1=CC(OC)=C(C(C2=C3C=NC=C2)=CN(C)C3=O)C=C1OC
Molecular Weight (g/mol) 353.41
Formula Weight 353.41
Percent Purity 99.86%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.