Learn More
GSK-872, MedChemExpress
Product Code. 16472796
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy
Product Code. 16472796
Brand: Medchem Express HY101872
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
MedChemExpress GSK-872 is a RIPK3 inhibitor, which binds RIP3 kinase domain with an IC50 of 1.8 nM, and inhibits kinase activity with an IC50 of 1.3 nM. GSK-872 decreases the RIPK3-mediated necroptosis and subsequent cytoplasmic translocation and expression of HMGB1, as well as ameliorates brain edema and neurological deficits in early brain injury.
GSK-872 is a RIPK3 inhibitor, which binds RIP3 kinase domain with an IC50 of 1.8 nM, and inhibits kinase activity with an IC50 of 1.3 nM. GSK-872 decreases the RIPK3-mediated necroptosis and subsequent cytoplasmic translocation and expression of HMGB1, as well as ameliorates brain edema and neurological deficits in early brain injury.
Specifications
Chemical Name or Material GSK-872
CAS 1346546-69-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H17N3O2S2
Quantity 25 mg
Solubility Information DMSO : 100 mg/mL (260.76 mM; Need ultrasonic)
SMILES O=S(C1=CC=C2N=CC=C(C2=C1)NC3=CC=C4SC=NC4=C3)(C(C)C)=O
Molecular Weight (g/mol) 383.49
Formula Weight 383.49
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.