Learn More
MedChemExpress IL-6 Protein, Human
Specifications
Specifications
| Content And Storage | Stored at -20°C. After reconstitution, it is stable at 4°C for 1 week or -20°C for longer. It is recommended to freeze aliquots at -20°C or -80°C for extended storage. |
| Quantity | 10 μg |
| Shipping Condition | Room temperature |
| Molecular Weight (g/mol) | Approximately 21,000 Da |
| Synonym | rHuIL-6∣ BSF-2∣ CDF∣ Hybridoma growth factor∣ IFN-beta-2 |