Learn More
ML241 hydrochloride, MedChemExpress
Product Code. 16443857
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy
Product Code. 16443857
Brand: Medchem Express HY19797A
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
MedChemExpress ML241 hydrochloride is a potent p97 inhibitor, inhibiting p97 ATPase with IC50 value of 100 nM.
ML241 hydrochloride is a potent p97 inhibitor, inhibiting p97 ATPase with IC50 value of 100 nM.
Specifications
Chemical Name or Material ML241 hydrochloride
CAS 2070015-13-1
Color White
Purity Grade Notes Research
Molecular Formula C23H25ClN4O
Quantity 50 mg
Solubility Information DMSO : ≥ 34 mg/mL (83.15 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES Cl[H].C1(N2C3=CC=CC=C3OCC2)=NC(NCC4=CC=CC=C4)=C5CCCCC5=N1
Molecular Weight (g/mol) 408.92
Formula Weight 408.92
Percent Purity 99.78%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.