Learn More
ML-098, MedChemExpress
Product Code. 16443696
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy
Product Code. 16443696
Brand: Medchem Express HY19800
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
MedChemExpress ML-098 (CID-7345532) is an activator of the GTP-binding protein Rab7 with an EC50 of 77.6 nM.
ML-098 (CID-7345532) is an activator of the GTP-binding protein Rab7 with an EC50 of 77.6 nM.
Specifications
Chemical Name or Material ML-098
CAS 878978-76-8
Color White
Purity Grade Notes Research
Molecular Formula C19H19NO3
Quantity 25 mg
Synonym CID-7345532
Solubility Information DMSO : ≥ 29 mg/mL (93.74 mM)
SMILES O=C(C1=CN(CCOC2=CC(C)=CC=C2C)C3=C1C=CC=C3)O
Molecular Weight (g/mol) 309.36
Formula Weight 309.36
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.