Learn More
MMAF-OMe, MedChemExpress
Description
MMAF-Ome, an antitubulin agent, is also an ADC cytotoxin. MMAF-Ome inhibits several tumor cell lines with IC50s of 0.056 nM, 0.166 nM, 0.183 nM, and 0.449 nM for MDAMB435/5T4, MDAMB361DYT2, MDAMB468, and Raji (5T4-) cell lines, respectively.
Specifications
Specifications
| Chemical Name or Material | MMAF-OMe |
| CAS | 863971-12-4 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C40H67N5O8 |
| Quantity | 2 mg |
| Synonym | Monomethyl auristatin F methyl ester |
| Solubility Information | DMSO : ≥ 100 mg/mL (134.05 mM) |
| SMILES | CC[C@H](C)[C@@H]([C@@H](CC(N1[C@@H](CCC1)[C@@H]([C@@H](C)C(N[C@H](C(OC)=O)CC2=CC=CC=C2)=O)OC)=O)OC)N(C([C@H](C(C)C)NC([C@H](C(C)C)NC)=O)=O)C |
| Molecular Weight (g/mol) | 745.99 |
| Show More |
Research purposes only