Learn More
Eppendorf™ CryoStorage Vials, 3D SafeCode
Product Code. 16416879 Shop All Eppendorf Products
Change view
Click to view available options
Volume (Metric):
0.5 mL
1.0 mL
1.5 mL
2.0 mL
4.0 mL
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 480
Pack of 960
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Sterility unitSize
16416879 4.0 mL Non-sterile Pack of 480
16476869 0.5 mL Non-sterile Pack of 960
16486869 1.0 mL Non-sterile Pack of 960
16496869 1.5 mL Non-sterile Pack of 480
16406879 2.0 mL Non-sterile Pack of 480
16426879 0.5 mL Sterile Pack of 960
16436879 1.0 mL Sterile Pack of 960
16446879 1.5 mL Sterile Pack of 480
16456879 2.0 mL Sterile Pack of 480
16466879 4.0 mL Sterile Pack of 480
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 16416879 Supplier Eppendorf™ Supplier No. 0030079442
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Provides a 3-level coding of the SafeCode system (datamatrix code, barcode, and clear writing) to enable safe sample identification. Up to 96 pre-labeled vials are pre-racked and pre-capped (0.5 mL to 4.0 mL) for storage applications.
  • 3-level coding for safe sample identification
  • ECC 200 error correction function enables safe code reading with up to 30 % damaged codes
  • Temperature range: -196 °C (gas phase of LN2) to +121 °C
  • Manufactured from high-quality polypropylene in class VIII cleanroom for highest sample purity
  • Pre-capped and pre-racked for comfortable handling
  • Additional information on vials available via Eppendorf website for documentation purposes
  • Direct import into Eppendorf eLABInventory and eLABJournal software for convenient handling
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Sterility Non-sterile
Volume (Metric) 4.0 mL
No. per Pack 48
RNase Free Yes
Dnase Free Yes
Barcode 2D SafeCode
Quantity 480 (10 Racks of 48 Vials)
Product Type Coded Cryobank Vial System