Learn More
Abnova™ Human TNFAIP2 Partial ORF (NP_006282, 552 a.a. - 650 a.a.) Recombinant Protein with GST-tag at N-terminal
Product Code. 16196785
Change view
Click to view available options
Quantity:
10 ug
25 ug
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16196785 25 ug 25µg
16186785 10 ug 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16196785 Supplier Abnova™ Supplier No. H00007127Q01.25ug
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Used for AP, Array, ELISA, WB-Re
This gene was identified as a gene whose expression can be induced by the tumor necrosis factor alpha (TNF) in umbilical vein endothelial cells. The expression of this gene was shown to be induced by retinoic acid in a cell line expressing a oncogenic version of the retinoic acid receptor alpha fusion protein, which suggested that this gene may be a retinoic acid target gene in acute promyelocytic leukemia. [provided by RefSeq]
Sequence: YILANADTIQHFCTQHGSPATWLQPALPTLAEIIRLQDPSAIKIEVATYATCYPDFSKGHLSAILAIKGNLSNSEVKRIRSILDVSMGAQEPSRPLFSL
Specifications
Accession Number NP_006282
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 7127
Molecular Weight (g/mol) 36.63kDa
Name TNFAIP2 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 ug
Immunogen YILANADTIQHFCTQHGSPATWLQPALPTLAEIIRLQDPSAIKIEVATYATCYPDFSKGHLSAILAIKGNLSNSEVKRIRSILDVSMGAQEPSRPLFSL
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias B94
Common Name TNFAIP2
Gene Symbol TNFAIP2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid