Learn More
Abnova™ Human C19orf46 Full-length ORF (AAH52573.1, 1 a.a. - 291 a.a.) Recombinant Protein with GST-tag at N-terminal
Product Code. 16178963
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
This item is not returnable. View return policy
Product Code. 16178963
Brand: Abnova™ H00163183P01.10ug
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Used for AP, Array, ELISA, WB-Re
Sequence: MALSLPLGPRLGSEPLNHPPGAPREADIVGCTVCPASGEESTSPEQAQTLGQDSLGPPEHFQGGPRGNEPAAHPPRWSTPSSYEDPAGGKHCEVFEEANTLDQDLEVEGDSDWPGPGGVWGPWAPSSLPTSTELEWDPAGDIGGLGPLGQKTARTLGVPCELCGHRGPQGRGQGLEEADTSHSRQDMLESGLGHQKRLARHQRHSLLRKPQDKKRQASPHLQDVRLEGNPGAPDPASRQPLTFLLILFLLFLLLVGAMFLLPASGGPCCSHARIPRTPYLVLSYVNGLPPV
Specifications
Accession Number AAH52573.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 163183
Molecular Weight (g/mol) 57.4kDa
Name C19orf46 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 μg
Immunogen MALSLPLGPRLGSEPLNHPPGAPREADIVGCTVCPASGEESTSPEQAQTLGQDSLGPPEHFQGGPRGNEPAAHPPRWSTPSSYEDPAGGKHCEVFEEANTLDQDLEVEGDSDWPGPGGVWGPWAPSSLPTSTELEWDPAGDIGGLGPLGQKTARTLGVPCELCGHRGPQGRGQGLEEADTSHSRQDMLESGLGHQKRLARHQRHSLLRKPQDKKRQASPHLQDVRLEGNPGAPDPASRQPLTFLLILFLLFLLLVGAMFLLPASGGPCCSHARIPRTPYLVLSYVNGLPPV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FLJ36445
Common Name C19orf46
Gene Symbol C19orf46
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid