Learn More
TfR (Transferrin R) Mouse anti-Human, Alexa Fluor 350, Clone: 29806, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB2474U-100UG
This item is not returnable.
View return policy
Description
TfR (Transferrin R) Monoclonal specifically detects TfR (Transferrin R) in Human samples. It is validated for Flow Cytometry.Specifications
| TfR (Transferrin R) | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| TFRC | |
| Human TfR | |
| RUO | |
| 7037 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 29806 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| CD71, CD71 antigen, CD71TRFR, p90, T9, TfR, TfR (TransferrinR), TFR1, TFRC, TR, transferrin receptor (p90, CD71), transferrin receptor protein 1, Trfr | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human TfR in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only (RUO)