Learn More
Gibco™ CD BHK-21 Production Medium
Product Code. 15349245
Click to view available options
Quantity:
1 x 100 L
1 x 10 L
10 kg
Unit Size:
0.2g
100 L
10kg
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
Product Code. 15349245
Brand: Gibco™ A1627701
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
CD BHK-21 Production Medium
Gibco™ CD BHK-21 Production Medium is a chemically defined, protein-free cell culture medium. It is designed to support the growth of BHK-21 cells in serum-free suspension culture. This medium is suitable for large-scale manufacturing applications.
CD BHK-21 Production Medium is manufactured as follows:
With
  • L-glutamine
  • Dry powder format
Without
  • Phenol Red
  • Protein hydrolysates
Gibco™ CD BHK-21 Production Medium is the first cell culture medium supporting complete serum-free and protein-free processes for BHK-21 cells. The dry powder format is easy to reconstitute, since it has no hydrolysates in the formulation. Adapting cells from serum-containing media is a rapid and simple procedure. BHK-21 cells can achieve high cell densities and maintain high cell viability in this medium.
Gibco™ CD BHK-21 Production Medium is suitable for large-scale suspension culture. Because it does not require supplementation with serum or other undefined components, Gibco™ CD BHK-21 Production Medium eliminates the risks associated with serum, while allowing you to achieve consistent virus titers and simplifying downstream purification.
cGMP Manufacturing
Gibco™ CD BHK-21 Production Medium is manufactured at a cGMP-compliant facility located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards.
TRUSTED_SUSTAINABILITY
Specifications
Cell Line BHK-21
Form Powder
Product Type Production Medium
With Additives Glutamine
Without Additives No Phenol Red
For Use With (Application) Viral Vaccine Production
Quantity 1 x 10 L
Content And Storage Store in refrigerator (2–8°C).
Culture Type Suspension Cell Culture
For Research Use or Further Manufacturing. Not for use in diagnostic procedures.