Learn More
Thermo Scientific™ Quantech™ Fluorometer Narrow-Band Excitation and Emission Filters
Product Code. 11771111 Shop All Thermo Scientific Products
Change view
Click to view available options
Wavelength:
254 nm
297 nm
313 nm
360 nm
365 nm
390 nm
405 nm
420 nm
440 nm
450 nm
460 nm
490 nm
520 nm
540 nm
577 nm
590 nm
680 nm
Unit Size:
Each
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Wavelength unitSize
11771111 540 nm Each
11721111 254 nm Each
11731111 440 nm Each
11741111 450 nm Each
11751111 460 nm Each
11651359 490 nm Each
11761111 520 nm Each
11781111 590 nm Each
11791111 680 nm Each
11701121 297 nm Each
11711121 360 nm Each
10324110 577 nm Each
10739978 313 nm Each
11771121 365 nm Each
11781121 390 nm Each
11791121 405 nm Each
11701131 420 nm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
This item is not returnable. View return policy
Product Code. 11771111 Supplier Thermo Scientific™ Supplier No. LE1095X15
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Thermo Scientific™ Quantech Digital Filter Fluorometer Narrow Band Filters allow you to choose the best excitation and emission wavelength ranges for your fluorescence experiment.
Thermo Scientific™ Quantech Digital Filter Fluorometer Narrow Band Filters allow you to choose the best excitation and emission wavelength ranges for your fluorescence experiment. Narrow band filters allow a narrow range of light to pass through the filter -- delivering the selectivity you need for accurate fluorescence assays.
Narrow band filters can be used to excite fluorescence or to measure a very small window of fluorescence emission. To select the proper filter for your assay, use the following guidelines:
  • There should be no overlap between the Narrow Band excitation filter and the Sharp Cut emission filter
  • A portion of the fluorescence excitation (absorption) spectrum near the peak falls in the wavelength range transmitted by the Narrow Band excitation filter
For the most sensitive measurement of fluorescence, an emission filter that does not overlap with this excitation filter must be selected.
TRUSTED_SUSTAINABILITY
Specifications
Type Narrow Band Filter
Wavelength 540 nm
For Use With (Equipment) Quantech