Learn More
CPC™ Plastic Quick-Disconnect Fittings
Product Code. 11700629
Change view
Click to view available options
Diameter (Metric) Inner:
6.4 mm
9.5 mm
Product Type:
Hose Barb Non-valved Coupling Insert
Hose Barb Non-valved In-line Coupling Body
Hose Barb Non-valved In-line Coupling Insert
Material:
ABS
Polycarbonate
Polysulfone
Color:
Amber
Purple
White
Unit Size:
Pack of 250
Pack of 5
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Product Type Material Color unitSize
11700629 9.5 mm Hose Barb Non-valved In-line Coupling Body Polysulfone Amber Pack of 5
17385589 9.5 mm Hose Barb Non-valved In-line Coupling Insert ABS White Pack of 250
17729529 6.4 mm Hose Barb Non-valved In-line Coupling Body Polycarbonate Purple Pack of 250
11780029 6.4 mm Hose Barb Non-valved In-line Coupling Body Polysulfone Amber Pack of 5
11739979 6.4 mm Hose Barb Non-valved Coupling Insert Polysulfone Amber Pack of 5
17739529 6.4 mm Hose Barb Non-valved In-line Coupling Insert Polycarbonate Purple Pack of 250
16215172 9.5 mm Hose Barb Non-valved In-line Coupling Insert Polycarbonate Amber Pack of 5
11749979 9.5 mm Hose Barb Non-valved In-line Coupling Insert Polycarbonate Amber Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 11700629 Supplier CPC™ Supplier No. 3131122
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables tests.
Detach these fittings with a simple press of the thumb latch. CPC™ Plastic Quick-Disconnect Fittings are ideal for use with corrosive fluids. Polycarbonate and polysulfone materials pass all physicochemical, cytotoxicity, leachables and extractables
CPC Series couplings assure secure connections, and are easy to operate—even with gloved hands. They are reusable, yet economical enough to allow disposability.
  • Material: Medical-grade ABS, polycarbonate and polysulfone
  • Tubing Range: 1/4 and 3/8 in. I.D. (6.4 and 9.5mm)
  • Fow Rate: Cv∽5.5 (79.25) max.
  • Ergonomic thumb latch
  • Audible 'click' when connected
  • USP Class VI materials
  • Can be sterilized by autoclave, EtO, e-beam, or gamma
  • Coupling halves can be rotated when connected
  • Tapered barb
  • BSE/TSE free materials
  • Cell culture media packaging and transfer lines
  • Harvest and collection applications
  • Buffer solutions
  • Tube sets
  • Urology drainage/collection systems
  • Hypo/hyperthermia blankets
Specifications
Certifications/Compliance USP Class VI, ADCF
Autoclavable Autoclavable
Diameter (Metric) Inner 9.5 mm
Material Polysulfone
Overall Length (English) 1.1 in.
Overall Length (Metric) 28 mm
For Use With 9.5mm
For Use With (Application) Cell culture media packaging and transfer lines, harvest and collection applications
Min. Operating Temperature (English) -40°F
Min. Operating Temperature (Metric) -40°C
Height (Metric) 25 mm
Height (English) 0.96 in.
Color Amber
Max. Operating Temperature (English) 300°F
Max. Operating Temperature (Metric) 149°C
Product Type Hose Barb Non-valved In-line Coupling Body