Learn More
Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh
Product Code. 11577532 Shop All Fisherbrand Products
Change view
Click to view available options
Pore Size:
20 μm
25 μm
32 μm
38 μm
45 μm
53 μm
63 μm
75 μm
90 μm
1.00 mm
106 μm
1.18 mm
125 μm
1.40 mm
150 μm
1.70 mm
180 μm
2 mm
212 μm
2.36 mm
250 μm
2.80 mm
300 μm
3.35 mm
355 μm
4 mm
425 μm
500 μm
600 μm
710 μm
852 μm
Unit Size:
Each
31 product options available for selection
Product selection table with 31 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
11577532 2.80 mm Each
11537542 1.00 mm Each
11567552 106 μm Each
11557552 125 μm Each
11547552 150 μm Each
11597532 2 mm Each
11527552 212 μm Each
11517552 250 μm Each
11527562 38 μm Each
11587542 425 μm Each
11517562 45 μm Each
11507552 300 μm Each
11597023 4 mm Each
11577542 500 μm Each
11507562 53 μm Each
11567542 600 μm Each
11597552 63 μm Each
11557542 710 μm Each
11587552 75 μm Each
11547542 852 μm Each
11577552 90 μm Each
11557562 20 μm Each
11547562 25 μm Each
11537562 32 μm Each
11597542 355 μm Each
11567532 3.35 mm Each
11587532 2.36 mm Each
11537552 180 μm Each
11507542 1.70 mm Each
11517542 1.40 mm Each
11527542 1.18 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 31 options available.
31 options
Product Code. 11577532
Only null left
Add to basket
Add to basket
Made from Stainless Steel and available in a wide variesty of sizes. Fisherbrand™ Stainless Steel Test Sieve with Woven Wire Mesh has a 200mm diameter.
100 mm and 200 mm diameter test sieves with Stainless Steel frame and woven wire mesh. Supplied with Certificate of Conformity and record card. Additional mesh sizes and diameter sieves available - please contact Customer Service for details.
Specifications
Quantity 1 Each
Pore Size 2.80 mm
Type Sieve
Certifications/Compliance Certificate of Conformity, BS 410: Part 1: 2000, ISO 3310: Part 1: 2000
Material Stainless Steel
Includes Certificate of conformity and record card
Diameter (Metric) Outer 200 mm
Frame Material Stainless Steel
Mesh Material Woven Wire