Learn More
Fisherbrand™ Anodized Aluminum Blocks
Click to view available options
Holds:
12 x 15 mL Tubes
12 x 16 mm Tubes
20 x 10 mm Tubes
20 x 13 mm Tubes
24 x 0.5 mL Tubes
24 x 1.5 mL Tubes
24 x 2.0 mL Tubes
35 x 6 mm Tubes
48 x 0.2 mL Tubes
5 x 50 mL Tubes
6 x 20 mm Tubes
6 x 25 mm Tubes
96-well PCR Plate
Unit Size:
Each
Description
- Excellent heat transfer
- Anodized to resist thermal shock
- For holding test tubes for digital heaters
- Available to fit a range of tube counts and sizes
Order Info
Holds
Specifications
Specifications
| Description | Holds 24 x 2.0mL Tubes |
| For Use With (Equipment) | For AccuBlock Digital Dry Bath |
| Holds | 24 x 2.0 mL Tubes |
| Material | Anodized Aluminum |
| Type | Block |
| For Use With (Application) | For Test Tubes |