Learn More
Beskrivning
For processing high-solute samples
Centriprep™ centrifugal filters are disposable ultrafiltration devices used for purifying, concentrating, and desalting biological samples in the 2–15 mL volume range. These complete, ready-to-use ultrafiltration devices are designed for operation in most centrifuges that can accommodate 50 mL centrifuge tubes. They are easy to use and offer a high flow rate. Centriprep™ devices consist of a sample container with a twist-lock cap, a filtrate collector containing a low-adsorptive, Ultracel™ regenerated cellulose membrane and an air-seal cap for sample isolation.
Features & Benefits:
- For use with samples containing high levels of solutes
- Unique inverse flow mode of operation with large deadstop
- Fast sample processing
- Available with Ultracel™ membranes: (3, 10, 30 and 50 kDa NMWL)
- Fits standard swinging-bucket rotor for 50 mL tubes
Applications:Concentration and Purification of Particle-laden Solutions, Solutions with High Solute Concentration, Separation of Low MW Solutes from Fermentation Broths, Cell Culture Media and Lysates
Specifikationer
Specifikationer
| Disposable | Single-use |
| Material (Membrane) | Ultracel-YM Regenerated Cellulose |
| MWCO | 50 kDa |
| Sterility | Non-sterile |
| Filtration Area | 2.84 cm2 |
| Housing Material | Regenerated Cellulose (RC) |
| Quantity | 96Pack |
| Type | Centrifugal Filters |
| Volume (Metric) | 12 to 15 mL |
| Length (Metric) | 135 mm |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?