missing translation for 'onlineSavingsMsg'
Learn More

Vidarabine, MedChemExpress

Product Code. 16422086
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
This item is not returnable. View return policy

Product Code. 16422086

Brand: Medchem Express HYB0277

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vidarabine (Ara-A) an antiviral agent which is active against herpes simplex and varicella zoster viruses. Vidarabine has IC50s of 9.3 μg/ml for HSV-1 and 11.3 μg/ml for HSV-2. Vidarabine also has anti-orthopoxvirus activity.

Vidarabine (Ara-A) an antiviral agent which is active against herpes simplex and varicella zoster viruses. Vidarabine has IC50s of 9.3 μg/ml for HSV-1 and 11.3 μg/ml for HSV-2. Vidarabine also has anti-orthopoxvirus activity.

Specifications

Chemical Name or Material Vidarabine
Physical Form Solid
CAS 5536-17-4
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C10H13N5O4
For Use With (Application) COVID-19-anti-virus
Synonym Ara-A Adenine Arabinoside 9-β-D-Arabinofuranosyladenine
Solubility Information DMSO : 50 mg/mL (187.10 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.51 mM; Need ultrasonic)
SMILES O[C@@H]([C@@H]1O)[C@@H](O[C@@H]1CO)N2C(N=CN=C3N)=C3N=C2
Molecular Weight (g/mol) 267.24
Formula Weight 267.24
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.