Learn More
Vidarabine, MedChemExpress
Product Code. 16478206
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
Product Code. 16478206
Brand: Medchem Express HYB0277
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Vidarabine (Ara-A) an antiviral agent which is active against herpes simplex and varicella zoster viruses. Vidarabine has IC50s of 9.3 μg/ml for HSV-1 and 11.3 μg/ml for HSV-2. Vidarabine also has anti-orthopoxvirus activity.
Vidarabine (Ara-A) an antiviral agent which is active against herpes simplex and varicella zoster viruses. Vidarabine has IC50s of 9.3 μg/ml for HSV-1 and 11.3 μg/ml for HSV-2. Vidarabine also has anti-orthopoxvirus activity.
Specifications
Chemical Name or Material Vidarabine
Physical Form Solid
CAS 5536-17-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C10H13N5O4
For Use With (Application) COVID-19-anti-virus
Synonym Ara-A Adenine Arabinoside 9-β-D-Arabinofuranosyladenine
Solubility Information DMSO : 50 mg/mL (187.10 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (2.51 mM; Need ultrasonic)
SMILES O[C@@H]([C@@H]1O)[C@@H](O[C@@H]1CO)N2C(N=CN=C3N)=C3N=C2
Molecular Weight (g/mol) 267.24
Formula Weight 267.24
Percent Purity 98.0%
Grade Research