missing translation for 'onlineSavingsMsg'
Learn More

VI 16832, MedChemExpress

Product Code. 16407548
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16407548

Brand: Medchem Express HY18623

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VI 16832 is a broad spectrum Type I kinase inhibitor which can be used as an enrichment tool for the comparative expression analysis of protein kinases in different cancer cell lines.

VI 16832 is a broad spectrum Type I kinase inhibitor which can be used as an enrichment tool for the comparative expression analysis of protein kinases in different cancer cell lines.

Specifications

Chemical Name or Material VI 16832
CAS 1430218-51-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H25N5O2
Quantity 10 mg
Solubility Information DMSO : 33.33 mg/mL (85.14 mM; Need ultrasonic)
SMILES O=C1C=CC2=CN=C(NC3=CC=C(OCCN)C=C3)N=C2N1C4C(C5)CCC5C4
Molecular Weight (g/mol) 391.47
Formula Weight 391.47
Percent Purity 99.72%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.