Learn More
Tocris Bioscience™ VH 032 amide-PEG1-acid
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
VH 032 amide-PEG1-acid is a functionalized von-Hippel-Lindau (VHL) protein ligand for PROTAC™ research and development; incorporates an E3 ligase ligand plus a PEG1 linker with terminal carboxylic acid ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D. PROTAC™ is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | 3-(3-((-1-((2 S,4 R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)amino)-3-oxopropoxy)propanoic acid |
| CAS | 2172820-07-2 |
| Quantity | 25 mg |
| Target | Degrader Building Blocks |
| Molecular Formula | C28H38N4O7S |
| Purity | 0.95 |