missing translation for 'onlineSavingsMsg'
Learn More

VGX-1027, MedChemExpress

Product Code. 16496578
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496578 10 mg 10mg
16402677 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16496578 Supplier Medchem Express Supplier No. HY15507

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VGX-1027 is an orally active isoxazole compound that exhibits various immunomodulatory properties. VGX-1027 targets macrophages, reducing the production of the proinflammatory mediators TNF-α, IL-1β, IL-10. VGX-1027 has antidiabetogenic effects by limiting cytokine-mediated immunoinflammatory events.

VGX-1027 is an orally active isoxazole compound that exhibits various immunomodulatory properties. VGX-1027 targets macrophages, reducing the production of the proinflammatory mediators TNF-α, IL-1β, IL-10. VGX-1027 has antidiabetogenic effects by limiting cytokine-mediated immunoinflammatory events.

Specifications

Chemical Name or Material VGX-1027
Physical Form Solid
CAS 6501-72-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C11H11NO3
For Use With (Application) COVID-19-immunoregulation
Synonym GIT 27
Solubility Information DMSO : ≥ 56 mg/mL (272.89 mM)
SMILES O=C(O)CC1CC(C2=CC=CC=C2)=NO1
Molecular Weight (g/mol) 205.21
Formula Weight 205.21
Percent Purity 99.93%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.