missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Verzistobart Humanized Recombinant Human Monoclonal Antibody
Human Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA555025
This item is not returnable.
View return policy
Description
Compatible with Direct ELISA.
Verzistobart inhibits the enzyme tyrosine kinase, blocking signal transduction pathways involved in cell proliferation and survival.
Specifications
| Verzistobart Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Agenus; INCAGN-02390; INCAGN-2390 | |
| Recombinant Human CD366/HAVCR2/TIM-3. | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA | |
| 1 mg/mL | |
| PBS with no preservative | |
| Human | |
| Protein A/G | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction