Learn More
Scientific Industries SI™ Vertical Tube Holder
Product Code. 11321474
Click to view available options
Tube Capacity:
15 mL
50 mL
50 mL, 15 mL, microtubes
No. of Positions:
12
3
6
6 x 50 mL, 6 x 15 mL Tubes, 12 Microtubes
Unit Size:
Each
Product Code. 11321474
Brand: Scientific Industries SI™ 83041241
Please to purchase this item. Need a web account? Register with us today!
Requires no additional attachment for use. Scientific Industries SI™ Vertical Tube Holder snaps 15 to 50mL tubes and microtubes into place.
  • Holds tubes vertically
  • Snaps on to any Vortex-Genie 2 Mixer and the Vortex-Genie Pulse
  • Optimized for high speed vortexing a small number of tubes
  • No additional attachment necessary
Specifications
Tube Capacity 15 mL
For Use With (Equipment) Vortex-Genie 2, Vortex-Genie Pulse
No. of Positions 12