Learn More
Ansell™ VersaTouch™ 92-205 Series Single-Use White Nitrile Gloves
Product Code. 12677051 Shop All Ansell Products
Click to view available options
Size:
7
8
9
10
Unit Size:
Pack of 100
Product Code. 12677051
Brand: Ansell™ 92205080
Only null left
Add to basket
Add to basket
Improved manufacturing processes for longer wear, greater tear strength and better grip in dry, wet and fatty conditions. A thinner glove with enhanced fingertip flexibility and improved overall comfort.
  • Single use gloves
  • Improved manufacturing processes for longer wear, greater tear strength and better grip in dry, wet and fatty conditions
  • Thinner glove with enhanced fingertip flexibility and improved overall comfort
  • Textured fingertips provide good comfort and tactility for sensitive, precise handling
  • Latex-free: no risk of Type 1 allergies
  • Approved for food contact applications
  • Ideal for handling all fatty food
Applications:
  • Prepared meals
  • Fruit and vegetable processing
  • Dairy processing
  • Food distribution
  • Catering
TRUSTED_SUSTAINABILITY
Specifications
Accelerator Free No
Applications Industrial
AQL 1.5
Color White
Disposable Single-use
Exterior Finish Fully Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Nitrile Gloves
Size 8
Sterility Non-sterile
Thickness (English) 2.9 mil
Thickness (Metric) 0.07 mm
Model 922058
Series 92-205