Learn More
Elga™ Pre-Treatment Cartridge
Click to view available options
:
For Small Prima, Option and Medica
Description
- Pretreatment cartridge
- Helps to produce general laboratory grade water at better than single distilled quality
- Applications include glassware washing/rinsing, feed to ultra pure water systems, reagent make-up/dilution, buffer and media preparation, general chemistry, qualitative analyses and feed to environmental cabinets
Specifications
Specifications
| For Use With (Application) | Glassware washing/rinsing, feed to ultra pure water systems, reagent make-up/dilution, buffer and media preparation, general chemistry, qualitative analyses and feed to environmental cabinets |
| For Use With (Equipment) | Purelab™ Option-S Series Water Purification Systems |