Learn More
Cole-Parmer™ Venturi Miniature Vacuum Pump
Shop All Cole-Parmer Products
Click to view available options
Flow Rate:
8.5 L/min.
Unit Size:
Each
Description
- No moving parts ensures long life with low maintenance.
- Venturi pumps are air-powered, require no electrical power source or lubricant.
- Place these compact vacuum pumps close to application for fast response and minimal vacuum loss.
Specifications
Specifications
| Max. Vacuum | 100 Mbar |
| Material | Aluminum |
| Flow Rate | 8.5 L/min. |
| Pump Type | Vacuum System |