missing translation for 'onlineSavingsMsg'
Learn More

Veledimex, MedChemExpress

Product Code. 16419727
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16419727

Brand: Medchem Express HY16785

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Veledimex (INXN-1001), a synthetic analog of the insect molting hormone ecdysone, is an orally active activator ligand for a proprietary gene therapy promoter system. Veledimex can be used to activate certain genes using the ecdysone receptor (EcR)-based inducible gene regulation system, the RheoSwitch Therapeutic System (RTS). Veledimex can cross blood-brain barrier (BBB) in both orthotopic GL-261 mice and cynomolgus monkeys.

Veledimex (INXN-1001), a synthetic analog of the insect molting hormone ecdysone, is an orally active activator ligand for a proprietary gene therapy promoter system. Veledimex can be used to activate certain genes using the ecdysone receptor (EcR)-based inducible gene regulation system, the RheoSwitch Therapeutic System (RTS). Veledimex can cross blood-brain barrier (BBB) in both orthotopic GL-261 mice and cynomolgus monkeys.

Specifications

Chemical Name or Material Veledimex
CAS 1093130-72-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H38N2O3
Quantity 5 mg
Synonym INXN-1001 RG-115932
Solubility Information DMSO : 25 mg/mL (57.00 mM; Need ultrasonic)
SMILES O=C(NN(C(C1=CC(C)=CC(C)=C1)=O)[C@@H](C(C)(C)C)CCC)C2=CC=CC(OC)=C2CC
Molecular Weight (g/mol) 438.6
Formula Weight 438.6
Percent Purity 98.45%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.