missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VEGFC Monoclonal Antibody (D3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA550862
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family, is active in angiogenesis and endothelial cell growth, and can also affect the permeability of blood vessels. This secreted protein undergoes a complex proteolytic maturation, generating multiple processed forms which bind and activate VEGFR-3 receptors. Only the fully processed form can bind and activate VEGFR-2 receptors. This protein is structurally and functionally similar to vascular endothelial growth factor D.
Specifications
| VEGFC | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| O35757, P49767 | |
| VEGFC | |
| Recombinant Rat VEGFC protein, Ala108-Arg223 (Accession #O35757). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2b κ |
| Western Blot, Immunohistochemistry (Paraffin), Western Blot | |
| D3 | |
| Unconjugated | |
| VEGFC | |
| AW228853; Flt4 ligand; FLT4 ligand DHM; Flt4-L; LMPH1D; Vascular endothelial growth factor C; vascular endothelial growth factor C isoform 129; vascular endothelial growth factor C isoform 184; vascular endothelial growth factor-related protein; VEGF3; VEGFC; VEGF-C; VRP | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 114111, 7424 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction