missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VE-cadherin Mouse anti-Human, CoraLite 594, Clone: 4F9B10, Proteintech
Shop All Proteintech ProductsDescription
The VE-cadherin antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human VE-cadherin. This antibody recognizes human antigen. The VE-cadherin antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Specifications
| Antigen | VE-cadherin |
| Applications | Immunofluorescence |
| Classification | Monoclonal |
| Clone | 4F9B10 |
| Conjugate | CoraLite 594 |
| Host Species | Mouse |
| Quantity | 100 μL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 1003 |
| Target Species | Human |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?