missing translation for 'onlineSavingsMsg'
Learn More

VcMMAE, MedChemExpress

Product Code. p-200097317
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16486638

Brand: Medchem Express HY15575

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VcMMAE (mc-vc-PAB-MMAE) is a drug-linker conjugate for ADC with potent antitumor activity by using the anti-mitotic agent, monomethyl auristatin E (MMAE, a tubulin inhibitor), linked via the lysosomally cleavable dipeptide, valine-citrulline (vc).

VcMMAE (mc-vc-PAB-MMAE) is a drug-linker conjugate for ADC with potent antitumor activity by using the anti-mitotic agent, monomethyl auristatin E (MMAE, a tubulin inhibitor), linked via the lysosomally cleavable dipeptide, valine-citrulline (vc).

Specifications

Chemical Name or Material VcMMAE
CAS 646502-53-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C68H105N11O15
Quantity 10 mg
Synonym MC-Val-Cit-PAB-MMAE mc-vc-PAB-MMAE
Solubility Information DMSO : ≥ 54 mg/mL (41.01 mM)
SMILES CC(C)[C@@H](C(N[C@@H](CCCNC(N)=O)C(NC1=CC=C(COC(N(C)[C@@H](C(C)C)C(N[C@@H](C(C)C)C(N([C@@H]([C@@H](C)CC)[C@H](OC)CC(N2[C@@]([C@H](OC)[C@@H](C)C(N[C@H](C)[C@@H](O)C3=CC=CC=C3)=O)([H])CCC2)=O)C)=O)=O)=O)C=C1)=O)=O)NC(CCCCCN4C(C=CC4=O)=O)=O
Molecular Weight (g/mol) 1316.63
Formula Weight 1316.63
Percent Purity 97.09%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.