Learn More
VBY-825, MedChemExpress
Product Code. 16469477
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16469477
Brand: Medchem Express HY15958
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress VBY-825 is a novel, reversible cathepsin inhibitor with high potency against cathepsins B, L, S and V.
VBY-825 is a novel, reversible cathepsin inhibitor with high potency against cathepsins B, L, S and V.
Specifications
Chemical Name or Material VBY-825
CAS 1310340-58-9
Color White
Purity Grade Notes Research
Molecular Formula C23H29F4N3O5S
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (233.40 mM; Need ultrasonic)
SMILES CC[C@H](NC([C@@H](N[C@@H](C1=CC=C(F)C=C1)C(F)(F)F)CS(=O)(CC2CC2)=O)=O)C(C(NC3CC3)=O)=O
Molecular Weight (g/mol) 535.55
Formula Weight 535.55
Percent Purity 99.29%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease