Learn More
VAV2 Mouse anti-Human, Unconjugated, Clone: 3F11B5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67108-1-IG-20UL
Description
The VAV2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human VAV2. This antibody recognizes human, pig, mouse, rat antigen. The VAV2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| VAV2 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, ELISA | |
| 3F11B5 | |
| Unconjugated | |
| 20 μL | |
| 7410 | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.