Learn More
B Braun™ Vasofix™ Safety cannula
Description
- Requires no user activation — no button, twists or clicks
- Automatically covers needle tip upon needle withdrawal
- Cannot be bypassed
- Eliminates risk of inadvertent activation during handling
- Stays in place through disposal
- Easy puncture at a wide range of angles
- Minimum effort of catheter insertion
- Self-activating Safety Shield – covers needle tip automatically after use
- Simplicity – looks and feels like a standard cannula
Specifications
Specifications
| Color | White |
| Needle Gauge | 17 G |
| Diameter (Metric) Inner Needle | 1.5 mm |
| Length (Metric) | 45 mm |