Learn More
B Braun™ Vasofix™ Safety cannula
Product Code. 15476970
Change view
Click to view available options
Needle Gauge:
17 G
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Needle Gauge unitSize
15476970 17 G Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15476970 Supplier B Braun™ Supplier No. 4268156S01
Please to purchase this item. Need a web account? Register with us today!
Reduce needlestick injuries with Vasofix Safety Cannula. B Braun™ Vasofix™ Safety cannula is perfect for minimizing needle-stick injuries. The cannula has an integrated automatic safety shield, which protects the needle tip to prevent needle-stick injuries.
  • Requires no user activation — no button, twists or clicks
  • Automatically covers needle tip upon needle withdrawal
  • Cannot be bypassed
  • Eliminates risk of inadvertent activation during handling
  • Stays in place through disposal
  • Easy puncture at a wide range of angles
  • Minimum effort of catheter insertion
  • Self-activating Safety Shield – covers needle tip automatically after use
  • Simplicity – looks and feels like a standard cannula
Specifications
Color White
Needle Gauge 17 G
Diameter (Metric) Inner Needle 1.5 mm
Length (Metric) 45 mm