missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ Vasofix™ Braunule Catheter

Product Code. 10484265
Click to view available options
Needle Gauge:
17 G
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 10484265

Brand: B Braun™ 4268156B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Certifications/Compliance ISO standard 10555-1 / 5
Material FEP
Color White
Needle Gauge 17 G
Diameter (Metric) Outer 45 mm
Latex Free Yes
Length (Metric) 38 mm
Product Type Vasofix Braunüle Catheter
Quantity Ea.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.