missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ Vasco™ Sensitive Examination Gloves

Product Code. 10147311
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Pack of 100
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 10147311

Brand: B Braun™ 6067565

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Manufactured from powder-free natural latex. B Braun™ Vasco™ Sensitive Examination Gloves are non-sterile with synthetic lining and ambidextrous fit. Micro-roughened surface provides high tactility and grip.

Non-sterile examination gloves in highly elastic natural latex

  • Powder-free
  • With synthetic lining for easy donning
  • Free of thiuram
  • Ambidextrous fit
  • Rolled edge
  • Micro-roughened surface
  • High tactility and grip
  • AQL 1.5 (tightness)
  • Tested to ASTM 1671-07 (Viral penetration) and EN 473-3
  • In accordance with DIN EN 455

Specifications

Classification PPE Category III
Color Code White
Exterior Finish Micro-rough
Latex Free No
Material Latex
Powdered Powder Free
Product Type Examination Gloves
Size Large
Thickness (Metric) ≥0.08 mm
Finger Style Textured
For Use With (Application) Suitable for food processing
Glove Cuff Style Rolled
Inspected for Medical Use Yes
Quantity 100 Pk.
Tensile Strength ≥18 MPa before aging, ≥14 MPa after aging
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.