Learn More
B Braun™ Vasco™ Powdered Examination Glove
Catalog Number 10425011
Click to view available options
Size:
Small
Medium
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Warning: Gloves contain natural latex which may cause allergic reactions including anaphylactic reactions.
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Warning: Gloves contain natural latex which may cause allergic reactions including anaphylactic reactions.
Gain a better grip with powered quality gloves. B Braun™ Vasco™ Powdered Examination Glove Powdered Examination Gloves are made of highly elastic natural latex properties.
  • Unsterile, powdered
  • Microrough surface
  • Thiuramfrei
  • In white
  • Complies with DIN EN
  • Tested according to EN 374
  • Protective glove according to EN 420 and PSA 89/686/EEC, Cat III
  • Suitable for food processing
Color White
Latex Free No
Material Latex
Powdered Powdered
Size Small
Sterility Non-sterile
Glove Cuff Color White