Learn More
Varenicline Tartrate, TRC
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Specifications
Specifications
| Chemical Name or Material | Varenicline Tartrate |
| CAS | 375815-87-5 |
| Molecular Formula | C13 H13 N3 . C4 H6 O6 |
| Quantity | 250 mg |
| Synonym | 6,10-Methano-6H-pyrazino[2,3-h][3]benzazepine','7,8,9,10-tetrahydro-','(2R,3R)-2,3-dihydroxybutanedioate (1:1), CP 526555-18, Varenicline Tartrate |
| InChI Formula | InChI=1S/C13H13N3.C4H6O6/c1-2-16-13-5-11-9-3-8(6-14-7-9)10(11)4-12(13)15-1;5-1(3(7)8)2(6)4(9)10/h1-2,4-5,8-9,14H,3,6-7H2;1-2,5-6H,(H,7,8)(H,9,10)/t8-,9+;1-,2-/m.1/s1 |
| SMILES | O[C@H]([C@@H](O)C(=O)O)C(=O)O.C1NC[C@H]2C[C@@H]1c3cc4nccnc4cc23 |
| IUPAC Name | (2R,3R)-2,3-dihydroxybutanedioic acid;(1R,12S)-5,8,14-triazatetracyclo[10.3.1.02,11.04,9]hexadeca-2,4,6,8,10-pentaene |
| Molecular Weight (g/mol) | 361.35 |
| Formula Weight | 361.1274 |