missing translation for 'onlineSavingsMsg'
Learn More

Vandetanib, MedChemExpress

Product Code. 16404816
Encompass_Preferred
Click to view available options
Quantity:
25 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
25mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16404816

Brand: Medchem Express HY10260

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Vandetanib (D6474) is a potent, orally active inhibitor of VEGFR2/KDR tyrosine kinase activity (IC50=40 nM). Vandetanib also has activity versus the tyrosine kinase activity of VEGFR3/FLT4 (IC50=110 nM) and EGFR/HER1 (IC50=500 nM).

Vandetanib (D6474) is a potent, orally active inhibitor of VEGFR2/KDR tyrosine kinase activity (IC50=40 nM). Vandetanib also has activity versus the tyrosine kinase activity of VEGFR3/FLT4 (IC50=110 nM) and EGFR/HER1 (IC50=500 nM).

Specifications

Chemical Name or Material Vandetanib
CAS 443913-73-3
Color White
Purity Grade Notes Research
Molecular Formula C22H24BrFN4O2
Quantity 100 mg
Synonym ZD6474
Solubility Information DMSO : 27.5 mg/mL (57.85 mM; Need ultrasonic and warming)
SMILES FC1=CC(Br)=CC=C1NC2=NC=NC3=CC(OCC4CCN(CC4)C)=C(C=C23)OC
Molecular Weight (g/mol) 475.35
Formula Weight 475.35
Percent Purity 95.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.