missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VAM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582663
This item is not returnable.
View return policy
Description
Immunogen sequence: NEILEDITPL INVDENVAEL VGILKEPHFQ SLLEAHDIVA SKCYDSPPSS PEMNNSSINN QLLPVDAIRI LGIHKRAGEP LGVTFRVENN DLVIARIL Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 95%.
Members of the peripheral membrane-associated guanylate kinase (MAGUK) family function in tumor suppression and receptor clustering by forming multiprotein complexes containing distinct sets of transmembrane, cytoskeletal, and cytoplasmic signaling proteins. All MAGUKs contain a PDZ-SH3-GUK core and are divided into 4 subfamilies, DLG-like (see DLG1), ZO1-like (see TJP1), p55-like (see MPP1), and LIN2-like (see CASK), based on their size and the presence of additional domains. MPP6 is a member of the p55-like MAGUK subfamily (Tseng et al., 2001).
Specifications
| VAM1 | |
| Polyclonal | |
| Unconjugated | |
| MPP6 | |
| CH474011:EDL88200; Dlgh4; Dlgh4 protein; Fibroblast growth factor-inducible protein 15; MAGUK p55 subfamily member 6; MAGUK protein p55T; membrane palmitoylated protein 6; membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 6); membrane protein, palmitoylated 6 (MAGUK p55 subfamily member 6); Mpp6; P55t; P55T protein; Pals2; protein associated with Lin7 2; protein associated with Lin-7 2; VAM1; VAM-1; VELI-associated MAGUK 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51678 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NZW5 | |
| MPP6 | |
| Recombinant protein corresponding to Human MPP6. Recombinant protein control fragment (Product #RP-92644). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction