Learn More
Upchurch Scientific Valve micro splitter valve graduated, SSt needle1/16 inch O.D., 0.020 inch (0.51mm)I.D. 1.2µL
Brand: Upchurch Scientific 42719-03
Valve micro splitter valve graduated, SSt needle1/16 inch O.D., 0.020 inch (0.51mm)I.D. 1.2µL