Learn More
Gibertini Elettronica™ VADE 4 Steam Generator
Determine volatile acidity and total sulphurous anhydride (SO2) with this steam generator. Gibertini Elettronica™ VADE 4 Steam Generator is an accessory unit for DEE Distillers.
Brand: Gibertini Elettronica™ VADE-4
Additional Details : Weight : 9.00000kg
Description
- Electrically heated boiler
- Small size allows for instantaneous and continuous steam generation
- Reduced pressurized steam volume for increased safety
Specifications
Steam Generator | |
220 V | |
220 V (-15%/+10%), 15 A | |
DEE Distillers | |
130 mm |
15 A | |
405 mm | |
Determining volatile acidity and total sulphurous anhydride (SO2) | |
307 mm |