Learn More
Leybold Vacuum Hose with Flanges ISO-KF
Description
- Simple to fit and remove
- Reliable, leak tight
- With ISO-KF Flanges, sizes DN 10 to DN 50
- Stainless steel
Specifications
Specifications
| Material | Stainless Steel |
| Includes | Flanges ISO-KF |
| For Use With (Application) | Vacuum pumps |
| Length (Metric) | 250 mm |
| Size | DN 25 |