Learn More
BD Vacutainer™ Safety-Lok™ Blood Collection Set
Click to view available options
Needle Gauge:
23 G
25 G
Length (Metric) Tube:
178 mm
Color:
Dark Blue
Light Blue
Unit Size:
Pack of 200
Pack of 25
Pack of 50
Description
Safety-Lok™ blood collection sets are sterile, closed systems for venous blood collection. They can also be used for short-term infusions of up to two hours. These winged infusion sets include a hypodermic needle, color-coded wings, an integrated safety shield, small-bore tubing and a connector. Many also include a Luer adapter.
- Safety mechanism protects against needle injuries following successful venipuncture
- Safety shield pushes over the needle, surrounding it completely
- Safety mechanism engages irreversibly with an audible click over the needle
- Single-handed activation so attention remains focused on the patient and the puncture site
Specifications
Specifications
| Product Type | Blood Collection Set |
| Color | Dark Blue |
| Needle Gauge | 25 G |
| Sterility | Sterile |
| For Use With (Application) | Venous blood collection, Short-term infusions |
| Includes | With luer adapter, integrated safety shield |
| Latex Free | Yes |
| Length (Metric) Needle | 19 mm |
| Length (Metric) Tube | 178 mm |
| Quantity | 200 Cs. |