Learn More
VAChT/SLC18A3 Antibody (S6-38), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-59378AF488
Product Code. 18202552
  • 631.00 € / 100µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
VAChT/SLC18A3 Monoclonal specifically detects VAChT/SLC18A3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
VAChT/SLC18A3
Monoclonal
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
SLC18A3
Synthetic peptide amino acids 521-532 of human VAChT
100 μL
6572
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
S6-38
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
MGC12716, solute carrier family 18 (vesicular acetylcholine), member 3, Solute carrier family 18 member 3, solute carrier family 18, member 3, VAChT, VACHTvesicular acetylcholine transporter
Mouse
Protein G purified
Primary
Detects 56kDa.
Store at 4C in the dark.
IgG1