Learn More
Wheaton™ Vaccum Filter With 40/35 Inner Joint
Brand:  Wheaton™ 419460
1021.25 EUR valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 1.00000kg
Product Code. 11768799
  • 1075.00 € / Each
Estimated Shipment: 19-09-2024
to see stock.
Add to basket
Specifications
Large Sample Volumes, Liquids With A Heavy Particulate Load And Viscous Liquids When A High Flow Rate Is Required
Vaccum Filter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products